1-Methyl-3,4-bis(3-indolyl)maleimide ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3,4-Di(1H-indol-3-yl)-1-methyl-1H-pyrrole-2,5-dione, Bisindolylmaleimide V

ALX270053M005
ENZOALX270053M005EA 296 EUR
ENZOALX270053M005
1-Methyl-3,4-bis(3-indolyl)maleimide ≥98%
1-Methyl-3,4-bis(3-indolyl)maleimide

Negative control for the protein kinase C (PKC)-inhibitory activity.

Formel: C₂₁H₁₅N₃O₂
Molecular Weight: 341,37 g/mol
MDL: MFCD00236433
CAS-Nummer: 113963-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR