HA-100 dihydrochloride ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: HA-100, 5-(Piperazin-1-ylsulphonyl)isoquinoline

ALX270069M005
ENZOALX270069M005EA 132 EUR
ENZOALX270069M005
HA-100 dihydrochloride ≥98% (durch HPLC)
HA-100 dihydrochloride

Protein kinase inhibitor


Inhibitor of myosin light chain kinase, cAMP-dependent protein kinase (PKA) and protein kinase C (PKC).

Formel: C₁₃H₁₅N₃O₂S
Molecular Weight: 277,3421 g/mol
MDL: MFCD00209852
CAS-Nummer: 84468-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR