Milrinon ≥97% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
6-Methyl-5-(4-pyridyl)-2-pyridone-3-carbonitrile, 2-Methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile, 3-Cyano-6-methyl-5-(4-pyridyl)-2-pyridone
ALX270083M005
ENZOALX270083M005EA
153
EUR
ENZOALX270083M005
Milrinon ≥97% (durch TLC)
Milrinon
PDE3 inhibitor
Selective inhibitor of phosphodiesterase 3 (PDE3, IC50=0.3µM).
Formel:
C₁₂H₉N₃O Molecular Weight: 211,22 g/mol |
MDL:
MFCD00133539 CAS-Nummer: 78415-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...