Pentoxifylline ≥99%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
3,7-Dimethyl-1-(5-oxohexyl)xanthine, 1-(5-Oxohexyl)theobromine, 3,7-Dimethyl-1-(5-oxohexyl)-1H-purine-2,6(3H,7H)-dione, 3,7-Dihydro-3,7-dimethyl-1-(5-oxohexyl)-1H-purine-2,6-dione
ALX270112G005
ENZOALX270112G005EA
50.6
EUR
ENZOALX270112G005
Pentoxifylline ≥99%
Pentoxifylline
PDE inhibitor
Phosphodiesterase inhibitor. Blocks the synthesis of TNF-α. Improves blood flow by decreasing blood viscosity.
Isolated from Penicillium expansum.
Formel:
C₁₃H₁₈N₄O₃ |
MDL:
MFCD00063379 CAS-Nummer: 6493-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...