Chlorpromazin-hydrochlorid ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride

ALX270171G005
ENZOALX270171G005EA 84.7 EUR
ENZOALX270171G005
Chlorpromazin-hydrochlorid ≥98% (durch HPLC)
Chlorpromazine hydrochloride

Antipsychotic working on a variety of receptors in the CNS


Inhibits calmodulin stimulation of cyclic nucleotide phosphodiesterase. Dopamine antagonist. D2 antagonist. H1 antagonist. Inhibits TNF-alpha production. Potent PLA2 inhibitor. Inhibits nitric oxide synthase (NOS) in mouse brain and prevents lipopolysaccharide induction of NOS in mouse lung. Antipsychotic.

Formel: C₁₇H₁₉ClN₂S
Molecular Weight: 355.32 g/mol
MDL: MFCD00012654
CAS-Nummer: 69-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR