Toluol-α-sulfonylfluorid 99 GC-FID

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: PMSF, Phenylmethylsulfonylfluorid, Phenylmethansulfonylfluorid, Phenylmethansulfonsäurefluorid, α-Toluolsulfonylfluorid, α-Toluolsulfonsäurefluorid

ALX270184G005 ALX270184G025
ENZOALX270184G005EA 94.6 EUR
ENZOALX270184G005 ENZOALX270184G025
Toluol-α-sulfonylfluorid 99 GC-FID
Toluol-α-sulfonylfluorid

Irreversible inhibitor of serine proteases. Widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity. Spleen membranes used for CB2 studies do not require this pretreatment, as FAAH activity is absent in these cells. Inhibits also cysteine proteases like papain (reversible by DTT treatment, as well as internucleosomal DNA fragmentation in immature thymocytes. For a related, more stable inhibitor see AEBSF.


  • Inhibitor of serine proteases and FAAH


White to off-white needle crystals.


Soluble in 100% ethanol, methanol or isopropanol; insoluble in water.

Formel: C₇H₇FO₂S
Molecular Weight: 174.2 g/mol
Siedepunkt: 80…85 °C (0.3 mmHg)
Schmelzpunkt: 91…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007424
CAS-Nummer: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR