Clofibrat ≥98% (durch GC)
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
ALX270256G001
ENZOALX270256G001EA
136
EUR
ENZOALX270256G001
Clofibrat ≥98% (durch GC)
Clofibrate
PPARα activator.
Activates PPARα. Induces cytochrome P450 4A1 and 4A3 which catalise the omega-oxidation of fatty acids and prostaglandins.
Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).
Formel:
C₁₂H₁₅ClO₃ Molecular Weight: 242,70 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000615 CAS-Nummer: 637-07-0 UN: 3082 ADR: 9,III Merck Index: 14,02377 |
Spezifikation Testergebnisse
Purity | ≥98% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...