Carnosolsäure ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (4aR,10aS)-5,6-Dihydroxy-7-isopropyl-1,1-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic acid

ALX270264M010 ALX270264M050
ENZOALX270264M010EA 85.8 EUR
ENZOALX270264M010 ENZOALX270264M050
Carnosolsäure ≥95% (durch HPLC)
Carnosic acid

PPARgamma activator. Antioxidant.


Naturally occurring phenolic compound with antioxidant properties. Inhibits lipid peroxidation induced by NADH or NADPH oxidation. Peroxisome proliferator-activated receptor γ (PPARγ) activator. Anti-inflammatory. Antimicrobial.


Isolated from Rosmarinus officinalis.

Formel: C₂₀H₂₈O₄
Molecular Weight: 332.44 g/mol
MDL: MFCD02259459
CAS-Nummer: 3650-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR