6-Hydroxy-2,5,7,8-tetramethyl-2-chromancarbonsäure ≥96,5% (durch titrimetrische Analyse)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Trolox, 6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid

ALX270267M100
ENZOALX270267M100EA 51.7 EUR
ENZOALX270267M100
6-Hydroxy-2,5,7,8-tetramethyl-2-chromancarbonsäure ≥96,5% (durch titrimetrische Analyse)
6-Hydroxy-2,5,7,8-tetramethyl-2-chromancarbonsäure

Cell permeable derivative of vitamin E. Antioxidant. Inhibits peroxynitrite mediated tyrosine nitration and mitochondrial proton permeability. Prevents lipid peroxidation, cytotoxicity and apoptosis induced by cytochrome P450 2E1 in HEP G2 cells.


  • Cell permeable vitamin E derivative


White to off-white, pale yellow or pale green powder.


Soluble in 100% ethanol (25 mg/ml) or methanol.

Formel: C₁₄H₁₈O₄
Molecular Weight: 250,29 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00006846
CAS-Nummer: 53188-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR