SB202190 hydrochloride ≥97%

Lieferant: ENZO LIFE SCIENCES

ALX270268M001 ALX270268M005
ENZOALX270268M001EA 138 EUR
ENZOALX270268M001 ENZOALX270268M005
SB202190 hydrochloride ≥97%
SB202190 hydrochloride

p38 inhibitor


A water soluble form of the potent p38 (SAPK2a) inhibitor SB202190 (Prod. No. BML-EI294).

Formel: C₂₀H₁₄FN₃O·HCl
CAS-Nummer: 350228-36-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR