5-AIQ hydrochloride ≥97% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-Aminoisoquinolinone hydrochloride

ALX270285M005
ENZOALX270285M005EA 384 EUR
ENZOALX270285M005
5-AIQ hydrochloride ≥97% (durch HPLC)
5-AIQ hydrochloride

Water-soluble, potent inhibitor of poly(ADP-ribose) polymerase-1 (PARP-1).


  • PARP-1 inhibitor


Beige to brown powder.


Soluble in water (warm).

Formel: C₉H₈N₂O·HCl
Molecular Weight: 160,2:36,5 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 93117-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR