Ubiquinone-5 ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Coenzyme Q1

ALX270294M002
ENZOALX270294M002EA 117 EUR
ENZOALX270294M002
Ubiquinone-5 ≥95% (durch HPLC)
Ubiquinone-5

Counteracts inhibitory effects on mitochondrial permeability transition pore (MPTP) of decylubiquinone (Prod. No. BML-CM115) and ubiquinone-10 (Prod. No. ALX-270-295).

Formel: C₁₄H₁₈O₄
Molecular Weight: 250,3 g/mol
CAS-Nummer: 727-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR