Ubiquinone-10 ≥90% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
ALX270295M002
ENZOALX270295M002EA 161 EUR
ENZOALX270295M002
Ubiquinone-10 ≥90% (durch HPLC)
Ubiquinone-10
MPTP inhibitor
Inhibitor of the mitochondrial permeability transition pore (MPTP). This inhibitory effect can be counteracted by ubiquinone-5 (Prod. No. ALX-270-294).
CAS-Nummer: 606-06-4