Sirtinol ≥97% (durch NMR)

Lieferant: ENZO LIFE SCIENCES

ALX270308M001 ALX270308M005
ENZOALX270308M001EA 51.7 EUR
ENZOALX270308M001 ENZOALX270308M005
Sirtinol ≥97% (durch NMR)
Sirtinol

SIRT inhibitor


Specific cell permeable inhibitor of the sirtuin family of NAD-dependent deacetylases (ySir2: IC50=48µM; hSIRT1: IC50=60µM; hSIRT2: IC50=58µM) with no effect on human HDAC1. Reported to inhibit Sir2p transcriptional silencing activity in vivo (IC50=25µM) and NAD-dependent histone deacetylase activity of purified recombinant yeast Sir2p (IC50=70µM) and hSIRT2 (IC50=40µM) in vitro.

Formel: C₂₆H₂₂N₂O₂
MDL: MFCD00810186
CAS-Nummer: 410536-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR