Compound 56 ≥97% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
4-[(3-Bromophenyl)amino]-6,7-diethoxyquinazoline
ALX270316C500
ENZOALX270316C500EA
340
EUR
ENZOALX270316C500
Compound 56 ≥97% (durch HPLC)
Compound 56
Potent and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR) (IC50=6pM).
- EGFR inhibitor
White to off-white powder.
Soluble in DMSO (200mg/ml).
Formel:
C₁₈H₁₈BrN₃O₂ Molecular Weight: 388,3 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
171745-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...