2,2'-[2,2-Propandiylbis(4,1-phenylenoxymethylen)]dioxiran ≥85%

Lieferant: ENZO LIFE SCIENCES

Synonyme: Bisphenol A Diglycidyl Ether, 2,2-Bis(4-hydroxyphenyl)propane Diglycidyl Ether, DER 332, BADGE, D.E.R. 332 epoxy resin

ALX270353G025
ENZOALX270353G025EA 55 EUR
ENZOALX270353G025
2,2'-[2,2-Propandiylbis(4,1-phenylenoxymethylen)]dioxiran ≥85%
2,2-Bis(4-glycidyloxyphenyl)propane

PPARgamma inhibitor


  • Low melting temperature solid.


Antagonist of PPARgamma. Binds to PPARgamma with an apparent Kd of 100µM.

Formel: C₂₁H₂₄O₄
Molecular Weight: 340,42 g/mol
MDL: MFCD00080480
CAS-Nummer: 1675-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR