Indirubin ≥98% (durch 1H-NMR, HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Indigo Red, [2,3'-Biindolinylidene]-2',3-dione
ALX270361M001
ALX270361M005
ENZOALX270361M001EA
84.7
EUR
ENZOALX270361M001
ENZOALX270361M005
Indirubin ≥98% (durch 1H-NMR, HPLC)
Indirubin
Inhibitor of CDK and GSK
Inhibitor of cyclin-dependent kinases (CDK1/cyclin B (IC50=10µM), CDK2/cyclin A (IC50=2.2µM), CDK2/cyclin E (IC50=7.5µM), CDK4/cyclin D1 (IC50=12µM), CDK5/p35 (IC50=5.5µM)) and of GSK-3beta (IC50=600nM). Inhibits inflammatory reaction in delayed-type hypersensitivity in mice. Ligand of human aryl hydrocarbon receptor. Exhibits anti-cancer activity.
Synthetic.
Formel:
C₁₆H₁₀N₂O₂ |
MDL:
MFCD00221745 CAS-Nummer: 479-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...