Indirubin ≥98% (durch 1H-NMR, HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Indigo Red, [2,3'-Biindolinylidene]-2',3-dione

ALX270361M001 ALX270361M005
ENZOALX270361M001EA 84.7 EUR
ENZOALX270361M001 ENZOALX270361M005
Indirubin ≥98% (durch 1H-NMR, HPLC)
Indirubin

Inhibitor of CDK and GSK


Inhibitor of cyclin-dependent kinases (CDK1/cyclin B (IC50=10µM), CDK2/cyclin A (IC50=2.2µM), CDK2/cyclin E (IC50=7.5µM), CDK4/cyclin D1 (IC50=12µM), CDK5/p35 (IC50=5.5µM)) and of GSK-3beta (IC50=600nM). Inhibits inflammatory reaction in delayed-type hypersensitivity in mice. Ligand of human aryl hydrocarbon receptor. Exhibits anti-cancer activity.


Synthetic.

Formel: C₁₆H₁₀N₂O₂
MDL: MFCD00221745
CAS-Nummer: 479-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR