TIQ-A ≥96% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Thieno[2,3-c]isoquinolin-5-one
ALX270365M005
ENZOALX270365M005EA
352
EUR
ENZOALX270365M005
TIQ-A ≥96% (durch HPLC)
TIQ-A
Potent poly(ADP-ribose) polymerase-1 (PARP-1) inhibitor (IC50=450nM). Neuroprotectant.
- PARP-1 inhibitor
Soluble in DMSO.
Formel:
C₁₁H₇NOS Molecular Weight: 201.2 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
420849-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...