EB-47 dihydrochloride dihydrate
Lieferant: ENZO LIFE SCIENCES
ALX270383M005
ENZOALX270383M005EA
362
EUR
ENZOALX270383M005
EB-47 dihydrochloride dihydrate
EB-47 dihydrochloride dihydrate
PARP-1 inhibitor
Very potent and water soluble PARP-1 inhibitor (IC50=45nM, 100% inhibition at 200nM). Shows cytoprotective effects against oxidative damage in cells and in in vivo models of reperfusion injury and inflammation.