NU 2058
Lieferant: ENZO LIFE SCIENCES
Synonyme:
NU2058, O-Cyclohexylmethylguanine, 6-(Cyclohexylmethoxy)-9H-purin-2-amine
ALX270394M025
ENZOALX270394M025EA
506
EUR
ENZOALX270394M025
NU 2058
NU 2058
Competitive inhibitor of CDK1/cyclin B (IC50=5µM) and CDK2/cyclin A (IC50=12µM). Pharmacologically distinct from the relative specific inhibitor of CDK1/2 roscovitine.
- CDK inhibitor
Soluble in water or DMSO.
Formel:
C₁₂H₁₇N₅O Molecular Weight: 247,30 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD05664734 CAS-Nummer: 161058-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...