Olomoucine II ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 6-(2-Hydroxybenzylamino)-2(R)-[[1-hydroxymethyl)propyl]amino]-9-isopropylpurine

ALX270396M005
ENZOALX270396M005EA 201 EUR
ENZOALX270396M005
Olomoucine II ≥98% (durch HPLC)
Olomoucine II

Selective inhibitor of CDK1/cyclin B (IC50=20nM). Potent cytotoxin in tumor cells: IC50=5.3µM against MCF7 cells, IC50=11.1µM against K562 cells, IC50=3.0µM against CEM cells, IC50=6.3µM against HOS cells, and IC50=6.3µM against G361 cells.


  • CDK/cyclin inhibitor


White to off-white crystalline solid.


Soluble in DMSO, dimethyl formamide or 100% ethanol.

Formel: C₁₉H₂₆N₆O₂
Molecular Weight: 370,5 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 500735-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR