Indirubin-3’-(2,3-dihydroxypropyl)oximether
Lieferant: ENZO LIFE SCIENCES
Synonyme:
E804
ALX270425M001
ENZOALX270425M001EA
198
EUR
ENZOALX270425M001
Indirubin-3’-(2,3-dihydroxypropyl)oximether
Indirubin-3’-(2,3-dihydroxypropyl)oximether
Src kinase inhibitor
Inhibitor of c-Src (IC50=0.43µM) blocking downstream STAT3 signalling. Induces apoptosis in human cancer cells.