Smoothened agonist ≥95%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-Chloro-N-(trans-4-(methylamino)cyclohexyl)-N-(3-(pyridin-4-yl)benzyl)benzo[b]thiophene-2-carboxamide, 3-Chloro-N-[trans-4-(methylamino)cyclohexyl]-N-[3-(4-pyridinyl)benzyl]-1-benzothiophene-2-carboxamide, SAG

ALX270426M001
ENZOALX270426M001EA 327 EUR
ENZOALX270426M001
Smoothened agonist ≥95%
Smoothened agonist

Smoothened ligand


A potent cell-permeable benzothiophene compound that modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM). Shown to induce Hedgehog pathway activation (EC50= ~3 nM in NIH 3T3-derived Shh-LIGHT2 cells) and counteracts Cyclopamine (BML-GR334) inhibition of Smo. Reported to act as an activator at low concentrations and as an inhibitor at very high concentrations.

Formel: C₂₈H₂₈ClN₃OS
Molecular Weight: 490,062 g/mol
MDL: MFCD22581513
CAS-Nummer: 912545-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR