SC-560 ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-(4-Chlorophenyl)-1-(4-methoxyphenyl)-3-(trifluoromethyl)-1H-pyrazole

ALX270427M005
ENZOALX270427M005EA 104 EUR
ENZOALX270427M005
SC-560 ≥98%
SC-560

Selective COX-1 inhibitor (IC50=9nM). Shows 700-fold more selectivity over COX-2 (IC50=6.3µM). Inhibits COX-1-derived platelet thromboxane B2, gastric PGE2, and dermal PGE2 production. Orally active in rat.


  • COX inhibitor


White to off-white crystalline solid.


Soluble in 100% ethanol, DMSO or dimethyl formamide; sparingly soluble in aqueous buffers.

Formel: C₁₇H₁₂N₂OClF₃
Molecular Weight: 352,7 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02179214
CAS-Nummer: 188817-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR