Rockout ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-(4-Pyridyl)-1H-indole

ALX270429M005
ENZOALX270429M005EA 50.6 EUR
ENZOALX270429M005
Rockout ≥98% (durch HPLC)
Rockout

A cell permeable, selective, and ATP-competitive inhibitor of Rho kinase (ROCK) activity (IC50=25µM). Does not inhibit the activation of Rho nor does it affect the in vitro activities of MLCK, PKCα, and SAPK2a/p38α. Shown to be less potent than Y-27632.


  • ROCK inhibitor


Off-white to beige solid.


Soluble in DMSO or dimethyl formamide.

Formel: C₁₃H₁₀N₂
Molecular Weight: 194.2 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 7272-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR