1,1-Dimethylbiguanid Hydrochlorid ≥97%
Lieferant: Enzo Life Sciences
Antidiabetic agent
Metformin is an antidiabetic agent. Reduces the cardiovascular complications of diabetes. Does not induce hypoglycemia. Inhibits hepatic gluconeogenesis via activation of the LKB1/AMPK pathway. Has antiproliferative effects in cancer cell lines.
Formel:
C₄H₁₂ClN₅ |
MDL:
MFCD00012582 CAS-Nummer: 1115-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...