SANT-2 ≥95%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-(3-(1H-Benzo[d]imidazol-2-yl)-4-chlorophenyl)-3,4,5-triethoxybenzamide
ALX270436M001
ENZOALX270436M001EA
271
EUR
ENZOALX270436M001
SANT-2 ≥95%
SANT-2
Smoothened inhibitor
Cell permeable antagonist of the Smoothened (Smo) protein function. Inhibitor of SAG (Prod. No. ALX-270-426) activity. Chemical tool to study the Hedgehog (Hh) signalling pathway and to explore therapeutic uses for inhibitors of Smo.
Formel:
C₂₆H₂₆ClN₃O₄ Molecular Weight: 480 g/mol |
MDL:
MFCD01827636 CAS-Nummer: 329196-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...