4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline ≥97% (durch 1H-NMR)

Lieferant: ENZO LIFE SCIENCES

Synonyme: LY364947, 4-(3-(Pyridin-2-yl)-1H-pyrazol-4-yl)quinoline

ALX270448M005
ENZOALX270448M005EA 161 EUR
ENZOALX270448M005
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline ≥97% (durch 1H-NMR)
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline

Cell permeable, potent, selective and ATP-competitive inhibitor of TGF-β RI kinase (IC50=51nM). Displays ~15-fold greater selectivity over p38α MAPK (IC50=740nM). Inhibits TGF-β-dependent cellular growth (IC50=89nM in NIH 3T3 mouse fibroblasts) and transcription activation (IC50=47nM in mink lung cells). Suppresses invasion of MDA-MB-231 breast cancer cells in a matrigel invasion assay.


Yellow to brown solid.


Soluble in DMSO or methanol.

Formel: C₁₇H₁₂N₄
Molecular Weight: 272.31 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00800758
CAS-Nummer: 396129-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR