DR2313
Lieferant: ENZO LIFE SCIENCES
ALX270452M025
ENZOALX270452M025EA
324
EUR
ENZOALX270452M025
DR2313
DR2313
PARP inhibitor
Potent, water soluble competitive PARP inhibitor (IC50=0.20µM and 0.24µM for PARP-1 and PARP-2 respectively).
Formel:
C₈H₁₀N₂OS |
MDL:
MFCD08703132 CAS-Nummer: 284028-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...