Sphingolactone-24 ≥95%
Lieferant: ENZO LIFE SCIENCES
ALX270453M001
ENZOALX270453M001EA
332
EUR
ENZOALX270453M001
Sphingolactone-24 ≥95%
Sphingolactone-24
Selective and irreversible inhibitor of neutral sphingomyelinase (N-SMase).
- Sphingomyelinase inhibitor
White waxy solid.
Soluble in DMSO (10mg/ml).
Formel:
C₁₈H₂₉NO₄ Lagertemperatur: Gefrierschrank |
CAS-Nummer:
881177-99-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...