Compound 15e ≥99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
PI3K-α Inhibitor, 3-(4-Morpholinothieno[3,2-d]pyrimidin-2-yl)phenol
ALX270455M005
ENZOALX270455M005EA
330
EUR
ENZOALX270455M005
Compound 15e ≥99% (durch HPLC)
Compound 15e
PI3 kinase inhibitor
Potent and specific inhibitor of phosphoinositide 3-kinasep110alpha isoform (IC50=0.0020µM). p110beta isoform (IC50=0.016µM)p110gamma isoform (IC50=0.66µM)C2beta isoform (IC50=0.22µM). Inhibits proliferation of A375 melanoma cells (IC50=0.58µM). Inhibits proliferation of U87MG human glioblastoma cells (IC50=1.10.µM).Inhibits proliferation of A2780 human ovarian cells (IC50=0.27µM).
Formel:
C₁₆H₁₅N₃O₂S Molecular Weight: 313.4 g/mol |
CAS-Nummer:
371943-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...