PQ401 ≥98% (durch HPLC und UV-Methode)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1-(5-Chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea, N-(5-Chloro-2-methoxyphenyl)-N’-(2-methyl-4-quinolinyl)urea, IGF-IR Inhibitor

ALX270459M005
ENZOALX270459M005EA 207 EUR
ENZOALX270459M005
PQ401 ≥98% (durch HPLC und UV-Methode)
PQ401

Potent inhibitor of insulin-like growth factor I receptor (IGF-IR) signalling and breast cancer cell growth in culture and in vivo.


  • IGF-1 receptor inhibitor


Off-white solid.


Soluble in DMSO (10mg/ml) or methanol (7mg/ml).

Formel: C₁₈H₁₆ClN₃O₂
Molecular Weight: 341,8 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00160558
CAS-Nummer: 196868-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR