AS-604850 ≥98% (durch HPLC und UV-Methode)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-(2,2-Difluoro-benzo[1,3]dioxol-5-ylmethylene)-thiazolidine-2,4-dione, PI3 Kinase p110γ Inhibitor, PI3Kγ inhibitor, 5-((2,2-Difluorobenzo[d][1,3]dioxol-5-yl)methylene)thiazolidine-2,4-dione

ALX270461M005
ENZOALX270461M005EA 203 EUR
ENZOALX270461M005
AS-604850 ≥98% (durch HPLC und UV-Methode)
AS-604850

Potent, cell permeable and ATP-competitive inhibitor of phosphoinositide 3-kinase γ (PI(3)Kγ). Exhibits selectivity over other PI(3)K isoforms.

Formel: C₁₁H₅F₂NO₄S
MDL: MFCD19705528
CAS-Nummer: 648449-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR