AS-605240

Lieferant: ENZO LIFE SCIENCES

ALX270462M005
ENZOALX270462M005EA 143 EUR
ENZOALX270462M005
AS-605240
AS-605240

Potent, cell permeable and ATP-competitive inhibitor of phosphoinositide 3-kinase γ (PI(3)Kγ). Exhibits selectivity over other PI(3)K isoforms.

Formel: C₁₂H₇N₃O₂S
MDL: MFCD11114066
CAS-Nummer: 648450-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR