TGX-221 ≥98% (durch NMR)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
7-Methyl-2-morpholino-9-(1-(phenylamino)ethyl)-4H-pyrido[1,2-a]pyrimidin-4-one
ALX270464M001
ENZOALX270464M001EA
1110
EUR
ENZOALX270464M001
TGX-221 ≥98% (durch NMR)
TGX-221
PI3 kinase p110beta inhibitor
Potent, selective and cell permeable inhibitor of phosphoinositide 3-kinase (PI(3)K) p110β.
Formel:
C₂₁H₂₄N₄O₂ Molecular Weight: 364,5 g/mol |
MDL:
MFCD11113209 CAS-Nummer: 663619-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...