Fluvastatin sodium salt ≥98% (durch HPLC)

Lieferant: Enzo Life Sciences

ALX270466M010 ALX270466M050
ENZOALX270466M010EA 79.2 EUR
ENZOALX270466M010 ENZOALX270466M050
Fluvastatin sodium salt ≥98% (durch HPLC)
Fluvastatin sodium salt

Anticholesterol agent. HMG-CoA inhibitor.


Anticholesterol agent. HMG-CoA reductase inhibitor and antioxidant.


Synthetic.

Formel: C₂₄H₂₅FNNaO₄
MDL: MFCD00929076
CAS-Nummer: 93957-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR