Sorafenib (BAY 43-9006) ≥95% (durch HPLC)

Lieferant: Enzo Life Sciences

ALX270469M001 ALX270469M005 ALX270469M025
ENZOALX270469M001EA 101 EUR
ENZOALX270469M001 ENZOALX270469M005 ENZOALX270469M025
Sorafenib (BAY 43-9006) ≥95% (durch HPLC)
Sorafenib (BAY 43-9006)

Broad-spectrum kinase inhibitor


Broad-spectrum kinase inhibitor.


Synthetic.

Formel: C₂₁H₁₆ClF₃N₄O₃
MDL: MFCD06411450
CAS-Nummer: 284461-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR