BIX 01294 trihydrochloride, Hyrdrat ≥95% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-(1-Benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine trihydrochloride, 2-(Hexahydro-4-methyl-1H-1,4-diazepin-1-yl)-6,7-dimethoxy-N-[1-(phenylmethyl)-4-piperidinyl]-4-quinazolinamine Trihydrochloride, BIX-01294 3HCl
ALX270473M002
ENZOALX270473M002EA
327
EUR
ENZOALX270473M002
BIX 01294 trihydrochloride, Hyrdrat ≥95% (durch HPLC)
BIX 01294 trihydrochloride
Selective and cell permeable inhibitor of G9a histone methyltransferase.
- G9a histone methyltransferase inhibitor
White solid.
Soluble in DMSO (5mg/ml).
Formel:
C₂₈H₃₈N₆O₂·3HCl Molecular Weight: 600,03 g/mol (anhy) Lagertemperatur: Gefrierschrank |
MDL:
MFCD16618384 CAS-Nummer: 1392399-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...