UPF1035 ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-Benzoyloxy-3,4-dihydroisoquinolin-1(2H)-one

ALX270498M005
ENZOALX270498M005EA 316 EUR
ENZOALX270498M005
UPF1035 ≥95% (durch HPLC)
UPF1035

PARP-2 specific inhibitor with 60-fold selectivity for PARP-2 (IC50=0.15 ± 0.04μM) over PARP-1 (IC50=9.0 ± 0.7μM). Can be used for further characterization of PARP-2 in pathophysiological conditions.


  • PARP-2 inhibitor


Soluble in DMSO.

Formel: C₁₆H₁₁NO₃
Molecular Weight: 265,3 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 370872-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR