CK-666 ≥95%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
2-Fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]-benzamide, 2-Fluoro-N-(2-(2-methyl-1H-indol-3-yl)ethyl)benzamide
ALX270506M002
ENZOALX270506M002EA
81.4
EUR
ENZOALX270506M002
CK-666 ≥95%
CK-666
Cell permeable inhibitor human (IC50=4µM), fission yeast (IC50=5µM), S. cerevisiae (IC50=12µM) and bovine (IC50=17µM) Arp2/3 complex between Arp2 and Arp3. Inhibits Listeria motility (IC50=7µM).
Tan to yellow solid.
Soluble in DMSO (a minimum concentration of 5 mg/ml is recommended).
Formel:
C₁₈H₁₇FN₂O Molecular Weight: 296,3 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD03036271 CAS-Nummer: 442633-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...