KS-176

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(4-(2-hydroxyethyl)phenyl)-2-(4-nitrobenzamido)benzamide, KS176

ALX270512M002
ENZOALX270512M002EA 381 EUR
ENZOALX270512M002
KS-176
KS-176

BCRP inhibitor


A potent specific inhibitor of Breast Cancer Resistance Protein (BCRP) / ATP-binding cassette sub-family G member 2 (ABCG2).

IC50=1.39µM

Formel: C₂₂H₁₉N₃O₅
Molecular Weight: 405,40 g/mol
MDL: MFCD22666402
CAS-Nummer: 1253452-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR