1,2-Dilauroyl-sn-glycero-3-phosphocholine ≥99% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: DLPC, (R)-2,3-Bis(dodecanoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate

ALX300034M250
ENZOALX300034M250EA 167 EUR
ENZOALX300034M250
1,2-Dilauroyl-sn-glycero-3-phosphocholine ≥99% (durch TLC)
1,2-Dilauroyl-sn-glycero-3-phosphocholine

20 mg/ml soluble in chloroform:methanol:acetic acid:water (100:30:10:2.5); slightly soluble in methanol.

Formel: C₃₂H₆₄NO₈P
Molecular Weight: 621,837 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00066561
CAS-Nummer: 18194-25-7

Bestellen

Spezifikation Testergebnisse

Purity ≥99% (TLC)
Identity Determined by IR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR