α-Mannosyl ceramide ≥95% (durch TLC, NMR)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
α-ManCer
ALX306031M001
ENZOALX306031M001EA
465
EUR
ENZOALX306031M001
α-Mannosyl ceramide ≥95% (durch TLC, NMR)
α-Mannosyl ceramide
Potent stimulator of NKT cells.
Potent stimulator of natural killer T (NKT) cells expressing the invariant Vα19-Jα26 T cell receptor α chain (Vα19 NKT cells).
Soluble in pyridine or warm DMSO (4 mg/ml).
Spezifikation Testergebnisse
Purity | ≥95% (TLC and NMR) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...