D-myo-Inositol-1,3,4,5,6-pentakisphosphate pentapotassium salt ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Ins(1,3,4,5,6)P5 . 5K

ALX307037M001
ENZOALX307037M001EA 440 EUR
ENZOALX307037M001
D-myo-Inositol-1,3,4,5,6-pentakisphosphate pentapotassium salt ≥98% (durch HPLC)
D-myo-Inositol-1,3,4,5,6-pentakisphosphate pentapotassium salt

Hygroscopic and soluble in water.

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR, ³¹P-NMR and MS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR