Isoins(1,4,5)P3/PM (caged) ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: D-2,3-O-Isopropylidene-6-O-(2-nitro-4,5-dimethoxy)benzyl-myo-inositol 1,4,5-trisphosphate-hexakis(propionoxymethyl)ester
ALX307071C020
ENZOALX307071C020EA 644 EUR
ENZOALX307071C020
Isoins(1,4,5)P3/PM (caged) ≥98% (durch HPLC)
Isoins(1,4,5)P3/PM (caged)
Ins(1,4,5)P3 derivative with a cage on position 6. Light pulse to destroy cage ~360 nm.
Soluble in DMSO, dichloromethane or methanol.
Bestellen
Spezifikation Testergebnisse
Purity ≥98% by HPLC
Identity Determined by ¹H-NMR, ³¹P-NMR and MS