Curcumin (aus Curcuma longa) ≥98% (durch HPLC), hochrein

Lieferant: ENZO LIFE SCIENCES

ALX350028M010 ALX350028M050 ALX350028M250
ENZOALX350028M010EA 51.7 EUR
ENZOALX350028M010 ENZOALX350028M050 ENZOALX350028M250
Curcumin (aus Curcuma longa) ≥98% (durch HPLC), hochrein
Curcumin
Formel: C₂₁H₂₀O₆
MDL: MFCD00008365
CAS-Nummer: 458-37-7
EINECS: 207-280-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR