Microcystin-RR
Lieferant: ENZO LIFE SCIENCES
ALX350043C100 ALX350043C250 ALX350043C050 ALX350043C100 ALX350043C250 ALX350043C500 ALX350043M001
ALEXALX350043C100EA 0 EUR
ALEXALX350043C100 ALEXALX350043C250 ENZOALX350043C050 ENZOALX350043C100 ENZOALX350043C250 ENZOALX350043C500 ENZOALX350043M001
Microcystin-RR
Proteine und Peptide
PP2A inhibitor
Arg-Arg analog of microcystin-LR (Prod. No. ALX-350-012). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A).
Isolated from Microcystis aeruginosa.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb9f3e327536ee465993549b60863ccc73893029&vid=35c07ea1a8f8a5239f0cae5deda073334c52a55e&prid=web&clid=SAPDAM