Microcystin-RR
Lieferant: ENZO LIFE SCIENCES
ALX350043C100
ALX350043C250
ALX350043C050
ALX350043C100
ALX350043C250
ALX350043C500
ALX350043M001
ALEXALX350043C100EA
0
EUR
ALEXALX350043C100
ALEXALX350043C250
ENZOALX350043C050
ENZOALX350043C100
ENZOALX350043C250
ENZOALX350043C500
ENZOALX350043M001
Microcystin-RR
Proteine und Peptide
PP2A inhibitor
Arg-Arg analog of microcystin-LR (Prod. No. ALX-350-012). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A).
Isolated from Microcystis aeruginosa.