Ouabain octahydrate ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Strophanthin G, 3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-1,5,11α-14,19-penta-hydroxycard-20(22)-enolide

ALX350066M100
ENZOALX350066M100EA 50.6 EUR
ENZOALX350066M100
Ouabain octahydrate ≥98% (durch TLC)
Ouabain octahydrate

Na⁺/K⁺ ATPase inhibitor


A steroidal inhibitor of cardiac sodium pumps (Na+/K+ ATPase) which has been recognized as an endogenous adrenal hormone. In the CNS it can be used as a tool to induce in vivo excitotoxicity. Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens.

Formel: C₂₉H₄₄O₁₂.8H₂O
Molecular Weight: 584,65 g/mol (anhy)
Lagertemperatur: Kühlschrank
MDL: MFCD00149240
CAS-Nummer: 11018-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR