Croton oil

Lieferant: ENZO LIFE SCIENCES
Gefahr

ALX350089KI225
ENZOALX350089KI225EA 3840 EUR
ENZOALX350089KI225
Croton oil
Croton oil

Natural source of phorbol and phorbol esters.


Purified from Croton tiglium L.


Soluble in DMSO, 100% ethanol or acetone. Insoluble in cold water.

Lagertemperatur: Kühlschrank
MDL: MFCD00130880
CAS-Nummer: 8001-28-3
Merck Index: 14,02598

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR